Cladribine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol, CdA, 2-Chloro-2'-deoxyadenosine, NSC105014, RWJ 26251
CAS Number: 4291-63-8
MDL Number: MFCD00153939
Molecular Formula: C10H12ClN5O3
Molecular Weight: 285.69
Purity/Analysis Method:
Product Categories: Bioscience, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2-Chloro-2'-deoxyadenosine, 2-Chloro-9-(2-deoxy-beta-D-arabinofuranosyl)adenine, CdA
Formula:
C₁₀H₁₂ClN₅O₃ MW: 285.7 g/mol Melting Pt: 220 °C |
MDL Number:
MFCD00153939 CAS Number: 4291-63-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...