6-Carboxyfluorescein ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

C2478-100MG
TCC2478-100MGEA 135 USD
TCC2478-100MG
6-Carboxyfluorescein ≥97.0% (by HPLC)
6-Carboxyfluorescein

CAS Number: 3301-79-9
MDL Number: MFCD00036873
Molecular Formula: C21H12O7
Molecular Weight: 376.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Lambda max.: 492 nm (0.1mol/L NaOH sol.)


Product Categories: Bioscience, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 6-FAM

Formula: C₂₁H₁₂O₇
MW: 376.32 g/mol
MDL Number: MFCD00036873
CAS Number: 3301-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449644e935ce7eae7e18da61a523b5227b84a4f3&vid=449644e935ce7eae7e18da61a523b5227b84a4f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR