2-Chlorothiazole ≥98.0%

Supplier: TCI America

C2471-25G C2471-5G
TCC2471-25GEA 149.6 USD
TCC2471-25G TCC2471-5G
2-Chlorothiazole ≥98.0%
2-Chlorothiazole

CAS Number: 3034-52-4 MDL Number: MFCD00210701 Molecular Formula: C3H2ClNS Molecular Weight: 119.57 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 145 Flash Point (°C): 54 Specific Gravity (20/20): 1.40 Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Thiazolyl Chloride

Formula: C₃H₂ClNS
Boiling Pt: 145 °C
Density: 1.40 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00210701
CAS Number: 3034-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ed5d2d14bcff7701ddd28a1e966a6066b25e1ad&vid=8ed5d2d14bcff7701ddd28a1e966a6066b25e1ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR