Crystal violet for biochemical research

Supplier: TCI America

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

C2460-5G
TCC2460-5GEA 49.89 USD
TCC2460-5G
Crystal violet for biochemical research
Crystal Violet

[for Biochemical Research]
CAS Number: 548-62-9
MDL Number: MFCD00011750
Molecular Formula: C25H30ClN3
Molecular Weight: 407.99
Form: Crystal
Color: Yellow Green
Melting point (°C): 211


Other Product Names / Synonyms: Basic Violet 3, Hexamethylpararosaniline Chloride

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 211 °C
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faebb1841dbe0a5dc8f857d6869bfa297142c3ba&vid=6d00940fd8eef621129559c76c92cca74bf2d814&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR