4-Cyano-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

C2456-1G C2456-5G
TCC2456-1GEA 38 USD
TCC2456-1G TCC2456-5G
4-Cyano-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Cyano-3-fluorophenylboronic acid

CAS Number: 843663-18-3 MDL Number: MFCD03411549 Molecular Formula: C7H5BFNO2 Molecular Weight: 164.93 Form: Crystal Color: White


Other Product Names / Synonyms: 4-Cyano-3-fluorobenzeneboronic Acid

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
MDL Number: MFCD03411549
CAS Number: 843663-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8397839cb6844fd24073efeabcdeb87b89951157&vid=8397839cb6844fd24073efeabcdeb87b89951157&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR