[12]Cycloparaphenylene ≥90.0%
Supplier: TCI America
Synonyms:
[12]CPP
C2449-10MG
TCC2449-10MGEA
1293
USD
TCC2449-10MG
[12]Cycloparaphenylene ≥90.0%
[12]Cycloparaphenylene
CAS Number: 1092522-75-2
Molecular Formula: C72H48
Molecular Weight: 913.18
Purity/Analysis Method:
Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials), Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: [12]CPP
Formula:
C₇₂H₄₈ MW: 913.18 g/mol |
CAS Number:
1092522-75-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...