C60MC12 ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
C60-fused N-Methylpyrrolidine-m-C12-phenyl
C2415-100MG
TCC2415-100MGEA
466.6
USD
TCC2415-100MG
C60MC12 ≥97.0% (by HPLC)
C60MC12
CAS Number: 403483-19-2
Molecular Formula: C81H35N
Molecular Weight: 1022.18
Purity/Analysis Method:
Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: C60-fused N-Methylpyrrolidine-m-C12-phenyl
Formula:
C₈₁H₃₅N MW: 1022.18 g/mol |
CAS Number:
403483-19-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...