Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
(1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene)(chloro)gold, 1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride
C2405-200MG
TCC2405-200MGEA
105
USD
TCC2405-200MG
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) ≥98.0% (by total nitrogen basis)
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I)
CAS Number: 852445-83-1
MDL Number: MFCD09839143
Molecular Formula: C27H36AuClN2
Molecular Weight: 621.02
Purity/Analysis Method:
Formula:
C₂₇H₃₆AuClN₂ MW: 621.02 g/mol Melting Pt: 298 °C |
MDL Number:
MFCD09839143 CAS Number: 852445-83-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...