Cobalt(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

C2388-250G
TCC2388-250GEA 96.18 USD
TCC2388-250G
Cobalt(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Cobalt (II) chloride hexahydrate

CAS Number: 7791-13-1 MDL Number: MFCD00149652 Molecular Formula: Cl2Co Molecular Weight: 129.83 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 87

Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 87 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR