1-Chloronaphthalene ≥97.0%

Supplier: TCI America

C2310-5G
TCC2310-5GEA 88 USD
TCC2310-5G
1-Chloronaphthalene ≥97.0%
1-Chloronaphthalene

CAS Number: 90-13-1 MDL Number: MFCD00003874 Molecular Formula: C10H7Cl Molecular Weight: 162.62 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 263 Melting point (°C): -20 Flash Point (°C): 121 Specific Gravity (20/20): 1.20

Formula: C₁₀H₇Cl
Boiling Pt: 263 °C
Melting Pt: –20 °C
Density: 1.20 (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00003874
CAS Number: 90-13-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e586bf747a50aa0f4d10ca02aabe2c077cab493&vid=2e586bf747a50aa0f4d10ca02aabe2c077cab493&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR