6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2306-5G C2306-1G
TCC2306-5GEA 372.4 USD
TCC2306-5G TCC2306-1G
6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)
6-Chloro-7-deazapurine

CAS Number: 3680-69-1 MDL Number: MFCD01686865 Molecular Formula: C6H4ClN3 Molecular Weight: 153.57 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 188


Product Categories: Bioscience, Nucleobases and Their Analogs Other Product Names / Synonyms: 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine

Formula: C₆H₄ClN₃
Melting Pt: 188 °C
MDL Number: MFCD01686865
CAS Number: 3680-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb2390f1c724da01fa0eea93da9353c110a93807&vid=fb2390f1c724da01fa0eea93da9353c110a93807&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR