Chloroquine diphosphate salt ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 50-63-5
MDL Number: MFCD00069852
Molecular Formula: C18H26ClN3
Molecular Weight: 515.86
Purity/Analysis Method:
Product Categories: Bioscience, Reagents for Pain & Itch Models, Antiprotozoal Ingredients
Other Product Names / Synonyms: N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate
Formula:
C₁₈H₂₆ClN₃ Melting Pt: 199 °C |
MDL Number:
MFCD00069852 CAS Number: 50-63-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...