2-(4-Chlorophenyl)ethanol ≥98.0%

Supplier: TCI America

Synonyms: 4-Chlorophenethyl alcohol

C2233-25G C2233-5G
TCC2233-25GEA 50 USD
TCC2233-25G 76219-678
2-(4-Chlorophenyl)ethanol ≥98.0%
2-(4-Chlorophenyl)ethanol

CAS Number: 1875-88-3 MDL Number: MFCD00002899 Molecular Formula: C8H9ClO Molecular Weight: 156.61 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 153 Flash Point (°C): 113 Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 4-Chlorophenethyl Alcohol

Formula: C₈H₉ClO
Boiling Pt: 153 °C
Density: 1.18 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d9af8d5078eace583d437f5816e2a2a6eedff3&vid=b0d9af8d5078eace583d437f5816e2a2a6eedff3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR