2,2'-O-Cyclocytidine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride

C2207-1G C2207-5G
TCC2207-1GEA 49 USD
TCC2207-1G TCC2207-5G
2,2'-O-Cyclocytidine hydrochloride ≥98.0%
2,2'-O-Cyclocytidine hydrochloride

CAS Number: 10212-25-6 MDL Number: MFCD00012636 Molecular Formula: C9H11N3O4 Molecular Weight: 261.66 Purity/Analysis Method: 98.0% (HPLC,N) Form: Crystal Melting point (°C): 264 Specific rotation [a]20/D: -21 deg (C=2, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs Other Product Names / Synonyms: 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride, Ancitabine Hydrochloride

Formula: C₉H₁₁N₃O₄
MW: 261.66 g/mol
Melting Pt: 264 °C
MDL Number: MFCD00012636
CAS Number: 10212-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ea437721c07e8faf608bb126ce8f0530093fdd9&vid=1ea437721c07e8faf608bb126ce8f0530093fdd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR