Cesium iodide ≥99.0% (by titrimetric analysis)
Supplier: TCI America
C2205-25G
TCC2205-25GEA
80.6
USD
TCC2205-25G
Cesium iodide ≥99.0% (by titrimetric analysis)
Cesium iodide
CAS Number: 7789-17-5
MDL Number: MFCD00010962
Molecular Formula: CsI
Molecular Weight: 259.81
Purity/Analysis Method:
Formula:
CsI Melting Pt: 632 °C |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...