2-Chlorothioanisole ≥98.0% (by GC)

Supplier: TCI America

C2069-25G C2069-5G
TCC2069-25GEA 63 USD
TCC2069-25G TCC2069-5G
2-Chlorothioanisole ≥98.0% (by GC)
2-Chlorothioanisole

CAS Number: 17733-22-1 MDL Number: MFCD00040124 Molecular Formula: C7H7ClS Molecular Weight: 158.64 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 227 Specific Gravity (20/20): 1.24


Other Product Names / Synonyms: 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene

Formula: C₇H₇ClS
Boiling Pt: 227 °C
Density: 1.24 (20 °C)
MDL Number: MFCD00040124
CAS Number: 17733-22-1
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94c8278fe693ff52f520989112d5cd1a215b05b1&vid=94c8278fe693ff52f520989112d5cd1a215b05b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR