Chlorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

C1948-500G
TCC1948-500GEA 24 USD
TCC1948-500G
Chlorobenzene ≥98.0% (by GC)
Chlorobenzene

CAS Number: 108-90-7 MDL Number: MFCD00000530 Molecular Formula: C6H5Cl Molecular Weight: 112.56 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 132 Melting point (°C): -45 Flash Point (°C): 28 Specific Gravity (20/20): 1.11

Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 132 °C
Melting Pt: –45 °C
Density: 1.11 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00000530
CAS Number: 108-90-7
UN: 1134
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd859faf3e376b2cd44c776a5504615703e9ca31&vid=dd859faf3e376b2cd44c776a5504615703e9ca31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR