kappa-Carrageenan

Supplier: TCI America

C1804-500G C1804-25G
TCC1804-500GEA 337 USD
TCC1804-500G TCC1804-25G
kappa-Carrageenan
kappa-Carrageenan

CAS Number: 11114-20-8 MDL Number: MFCD00151514 Form: Crystal Color: White


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)

Formula: C₁₂H₁₇O₁₃S
MDL Number: MFCD00151514
CAS Number: 11114-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaf41182262a99d7e4bf5194088e3f9e5c1382dd&vid=eaf41182262a99d7e4bf5194088e3f9e5c1382dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR