kappa-Carrageenan
Supplier: TCI America
C1804-500G
C1804-25G
TCC1804-500GEA
337
USD
TCC1804-500G
TCC1804-25G
kappa-Carrageenan
kappa-Carrageenan
CAS Number: 11114-20-8
MDL Number: MFCD00151514
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
Formula:
C₁₂H₁₇O₁₃S |
MDL Number:
MFCD00151514 CAS Number: 11114-20-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...