3-Chlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1791-1G C1791-5G
TCC1791-1GEA 45 USD
TCC1791-1G TCC1791-5G
3-Chlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-Chlorobenzenesulphonyl chloride

CAS Number: 2888-06-4 MDL Number: MFCD00051697 Molecular Formula: C6H4Cl2O2S Molecular Weight: 211.06 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 135 Flash Point (°C): 110 Freezing point (°C): 25

Formula: C₆H₄Cl₂O₂S
Boiling Pt: 135 °C
Flash Pt: 110 °C
MDL Number: MFCD00051697
CAS Number: 2888-06-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52cf897833c12d65dfbc9c055a30ec52d9b34700&vid=52cf897833c12d65dfbc9c055a30ec52d9b34700&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR