Cytidine-5'-monophosphate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
5'-Cytidylic Acid, 5'-CMP, ((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
C1675-5G
C1675-1G
TCC1675-5GEA
166.42
USD
TCC1675-5G
TCC1675-1G
Cytidine-5'-monophosphate ≥98.0% (by HPLC)
Cytidine-5'-monophosphate
CAS Number: 63-37-6
MDL Number: MFCD00006544
Molecular Formula: C9H14N3O8P
Molecular Weight: 323.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 9.8 deg (C=1, 0.5mol/L Na2HPO4 sol.)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: 5'-Cytidylic Acid, 5'-CMP
Formula:
C₉H₁₄N₃O₈P |
MDL Number:
MFCD00006544 CAS Number: 63-37-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...