Cortexolone ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
17alpha,21-Dihydroxy-4-pregnene-3,20-dione, Reichstein's Substance S, 11-Deoxy-17-hydroxycorticosterone
C1478-1G
TCC1478-001GEA
238.49
USD
TCC1478-001G
Cortexolone ≥98.0% (by HPLC)
Cortexolone
CAS Number: 152-58-9
MDL Number: MFCD00003662
Molecular Formula: C21H30O4
Molecular Weight: 346.47
Purity/Analysis Method:
Product Categories: Bioscience, Pregnanes (Glucocorticoids), Steroids
Other Product Names / Synonyms: 11-Deoxy-17-hydroxycorticosterone, 17alpha,21-Dihydroxy-4-pregnene-3,20-dione, Reichstein's Substance S
Formula:
C₂₁H₃₀O₄ MW: 346.47 g/mol |
MDL Number:
MFCD00003662 CAS Number: 152-58-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...