Carbonylhydridotris(triphenylphosphine)rhodium(I)
Supplier: TCI America
Synonyms:
Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)
C1383-1G
C1383-5G
TCC1383-001GEA
397
USD
TCC1383-001G
TCC1383-5G
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Carbonylhydridotris(triphenylphosphine)rhodium(I)
CAS Number: 17185-29-4
MDL Number: MFCD00151644
Molecular Formula: C55H46OP3Rh
Molecular Weight: 918.80
Form: Crystal
Color: Yellow
Melting point (°C): 122
Other Product Names / Synonyms: Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride
Formula:
C₅₅H₄₆OP₃Rh MW: 918.80 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00151644 CAS Number: 17185-29-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...