4-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
C1353-5G
C1353-1G
TCC1353-005GEA
100
USD
TCC1353-005G
TCC1353-001G
4-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
4-Carboxyphenylboronic acid
CAS Number: 14047-29-1
MDL Number: MFCD00151801
Molecular Formula: C7H7BO4
Molecular Weight: 165.94
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-Carboxybenzeneboronic Acid
Formula:
C₇H₇BO₄ |
MDL Number:
MFCD00151801 CAS Number: 14047-29-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...