Allyl methyl carbonate ≥98.0%
Supplier: TCI America
CAS Number: 35466-83-2
MDL Number: MFCD00134545
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Carbonic Acid Allyl Methyl Ester
Formula:
C₅H₈O₃ Boiling Pt: 131 °C Density: 1.03 (20 °C) Flash Pt: 33 °C |
MDL Number:
MFCD00134545 CAS Number: 35466-83-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...