1,4-Dioxaspiro[4.5]decan-8-one ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

C1292-5G C1292-100G C1292-25G
TCC1292-005GEA 18 USD
TCC1292-005G TCC1292-100G TCC1292-025G
1,4-Dioxaspiro[4.5]decan-8-one ≥98.0%
1,4-Dioxaspiro[4.5]decan-8-one

CAS Number: 4746-97-8 MDL Number: MFCD00010214 Molecular Formula: C8H12O3 Molecular Weight: 156.18 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 73


Other Product Names / Synonyms: 1,4-Dioxaspiro[4.5]decan-8-one

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00010214
CAS Number: 4746-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58bd5515fa32c5b33ffa0619282fee0852fe6696&vid=58bd5515fa32c5b33ffa0619282fee0852fe6696&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR