1,4-Dioxaspiro[4.5]decan-8-one ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal
C1292-5G
C1292-100G
C1292-25G
TCC1292-005GEA
18
USD
TCC1292-005G
TCC1292-100G
TCC1292-025G
1,4-Dioxaspiro[4.5]decan-8-one ≥98.0%
1,4-Dioxaspiro[4.5]decan-8-one
CAS Number: 4746-97-8
MDL Number: MFCD00010214
Molecular Formula: C8H12O3
Molecular Weight: 156.18
Purity/Analysis Method:
Other Product Names / Synonyms: 1,4-Dioxaspiro[4.5]decan-8-one
Formula:
C₈H₁₂O₃ MW: 156.18 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD00010214 CAS Number: 4746-97-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...