2-Chloroethyl ethyl ether ≥98.0%

Supplier: TCI America

Synonyms: 2-Ethoxyethyl chloride, 2-Chlorodiethyl ether, 2-Chloroethylethylether

C1172-25ML C1172-500ML
TCC1172-025MLEA 93.1 USD
TCC1172-025ML TCC1172-500ML
2-Chloroethyl ethyl ether ≥98.0%
2-Chloroethyl ethyl ether

CAS Number: 628-34-2 MDL Number: MFCD00000974 Molecular Formula: C4H9ClO Molecular Weight: 108.57 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 108 Flash Point (°C): 6 Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-Ethoxyethyl Chloride

Formula: C₄H₉ClO
Boiling Pt: 108 °C
Melting Pt: –60 °C
Density: 1.00 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00000974
CAS Number: 628-34-2
EINECS: 211-037-9
UN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22a88b9e7e44c48d514f6cfc2b65422cb09b0dd3&vid=22a88b9e7e44c48d514f6cfc2b65422cb09b0dd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR