Phthalocyanine chloroaluminum ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Chloroaluminum Phthalocyanine
C1167-1G
TCC1167-001GEA
65
USD
TCC1167-001G
TCC1167-5G
Phthalocyanine chloroaluminum ≥98.0% (by titrimetric analysis)
Phthalocyanine chloroaluminum
CAS Number: 14154-42-8
MDL Number: MFCD00049386
Molecular Formula: C32H16AlClN8
Molecular Weight: 574.97
Purity/Analysis Method:
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Solar Cell Research Reagents, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Chloroaluminum Phthalocyanine
Formula:
C₃₂H₁₆AlClN₈ MW: 574.97 g/mol |
MDL Number:
MFCD00049386 CAS Number: 14154-42-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...