Chloroform ≥99.5% stabilized

Supplier: TCI America
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with Stabilised

C1111-500ML
TCC1111-500MLEA 32.11 USD
TCC1111-500ML
Chloroform ≥99.5% stabilized
Chloroform

(stabilized with 2-Methyl-2-butene) [for HPLC Solvent] CAS Number: 67-66-3 MDL Number: MFCD00000826 Molecular Formula: CHCl3 Molecular Weight: 119.37 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Boiling point (°C): 60 Melting point (°C): -64 Specific Gravity (20/20): 1.49

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 60 °C
Melting Pt: –64 °C
Density: 1.49 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86e586046aafe76663bc99306f37165bea84d15e&vid=d47a6061a43406057bac2f9ee2d9db2c075a8586&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR