2-Chloronicotinic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
C1108-250G
C1108-25G
TCC1108-250GEA
100
USD
TCC1108-250G
TCC1108-025G
2-Chloronicotinic acid ≥98.0%
2-Chloronicotinic acid
CAS Number: 2942-59-8
MDL Number: MFCD00006236
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Chloro-3-pyridinecarboxylic Acid
Formula:
C₆H₄ClNO₂ Boiling Pt: 284 °C (1013 hPa) Melting Pt: 181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006236 CAS Number: 2942-59-8 EINECS: 220-937-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...