5-Chloroindole ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-1H-indole

C1107-5G C1107-25G
TCC1107-005GEA 96 USD
TCC1107-005G TCC1107-025G
5-Chloroindole ≥98.0%
5-Chloroindole

CAS Number: 17422-32-1 MDL Number: MFCD00005672 Molecular Formula: C8H6ClN Molecular Weight: 151.59 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 130 Melting point (°C): 72

Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 130 °C
Melting Pt: 72 °C
MDL Number: MFCD00005672
CAS Number: 17422-32-1
EINECS: 241-448-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060b2e507efab53205d8b2076e08a722d2bc77ed&vid=060b2e507efab53205d8b2076e08a722d2bc77ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR