2-Chlorobenzylamine ≥98.0%

Supplier: TCI America

Synonyms: (2-Chlorophenyl)methanamine

C1065-25G
TCC1065-25GEA 33 USD
TCC1065-25G
2-Chlorobenzylamine ≥98.0%
2-Chlorobenzylamine

CAS Number: 89-97-4 MDL Number: MFCD00008108 Molecular Formula: C7H8ClN Molecular Weight: 141.60 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 219 Flash Point (°C): 88 Specific Gravity (20/20): 1.17

Formula: C₇H₈ClN
Boiling Pt: 219 °C
Density: 1.17 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00008108
CAS Number: 89-97-4
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07c166909f43c62c4e1f0fda13464449edce0bc8&vid=07c166909f43c62c4e1f0fda13464449edce0bc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR