2-Chlorobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (2-Chlorophenyl)methanol

C0973-25G
TCC0973-025GEA 48 USD
TCC0973-025G
2-Chlorobenzyl alcohol ≥98.0%
2-Chlorobenzyl alcohol

CAS Number: 17849-38-6 MDL Number: MFCD00004604 Molecular Formula: C7H7ClO Molecular Weight: 142.58 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 135 Melting point (°C): 71

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227
Melting Pt: 71 °C
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR