Cyclopentadienylthallium ≥95.0%

Supplier: TCI America

Synonyms: Thallium(I) cyclopentadienide

C0954-5G
TCC0954-005GEA 365 USD
TCC0954-005G
Cyclopentadienylthallium ≥95.0%
Cyclopentadienylthallium

CAS Number: 34822-90-7 MDL Number: MFCD00134141 Molecular Formula: C5H5Tl Molecular Weight: 269.48 Purity/Analysis Method: 95.0% (T) Form: Crystal

Formula: C₅H₅Tl
Boiling Pt: 90…110 °C (1 mmHg)
Melting Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134141
CAS Number: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90a7ff8986b3e836ee7b3fb77a0b180461c194dd&vid=90a7ff8986b3e836ee7b3fb77a0b180461c194dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR