Cyclopentyl phenyl ketone ≥98.0%

Supplier: TCI America

Synonyms: Cyclopentyl(phenyl)methanone, Benzoylcyclopentane

C0882-5ML
TCC0882-005MLEA 139 USD
TCC0882-005ML
Cyclopentyl phenyl ketone ≥98.0%
Cyclopentyl phenyl ketone

CAS Number: 5422-88-8 MDL Number: MFCD00001378 Molecular Formula: C12H14O Molecular Weight: 174.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 159 Flash Point (°C): 110 Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Benzoylcyclopentane

Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 159 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001378
CAS Number: 5422-88-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR