5-Chloro-2-pentanone ≥98.0% stabilised

Supplier: TCI America

Synonyms: 5-Chloropentan-2-one

C0867-25ML
TCC0867-25MLEA 186.2 USD
TCC0867-25ML
5-Chloro-2-pentanone ≥98.0% stabilised
5-Chloro-2-pentanone

(stabilized with K2CO3) CAS Number: 5891-21-4 MDL Number: MFCD00001008 Molecular Formula: C5H9ClO Molecular Weight: 120.58 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 106 Flash Point (°C): 35 Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 3-Chloropropyl Methyl Ketone

Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 71
Melting Pt: -48
Density: 1.05 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00001008
CAS Number: 5891-21-4
UN: 1224
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=693a9f528a2c01f3894773c701f8a04f58ef8d91&vid=693a9f528a2c01f3894773c701f8a04f58ef8d91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR