Chloroform ≥99.0% stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with Stabilised

C0819-500ML
TCC0819-500MLEA 34.91 USD
TCC0819-500ML
Chloroform ≥99.0% stabilized
Chloroform

(stabilized with Ethanol) [for HPLC Solvent] CAS Number: 67-66-3 MDL Number: MFCD00000826 Molecular Formula: CHCl3 Molecular Weight: 119.37 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 60 Melting point (°C): -64 Specific Gravity (20/20): 1.48

Formula: CHCl₃
Boiling Pt: 60 °C
Melting Pt: –64 °C
Density: 1.48 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=519d4115a883c19feb6df867776f36e2712aca37&vid=e5872f4a805b4f4e2adc0882e25118d498598eb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR