Capsanthin (=paprika extract) in vegetable oil solution
Supplier: TCI America
C0781-25G
TCC0781-025GEA
101.25
USD
TCC0781-025G
Capsanthin (=paprika extract) in vegetable oil solution
Capsanthin (=paprika extract)
CAS Number: 465-42-9
MDL Number: MFCD03423725
Molecular Formula: C40H56O3
Molecular Weight: 584.89
Form: Liquid
Color: Red
Other Product Names / Synonyms: Paprika Extract
Formula:
C₄₀H₅₆O₃ |
MDL Number:
MFCD03423725 CAS Number: 465-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...