Cyclohexane ≥99.5%

Supplier: TCI America
Danger

C0696-100ML
TCC0696-100MLEA 67.57 USD
TCC0696-100ML
Cyclohexane ≥99.5%
Cyclohexane

[for Spectrophotometry] CAS Number: 110-82-7 MDL Number: MFCD00003814 Molecular Formula: C6H12 Molecular Weight: 84.16 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Boiling point (°C): 81 Flash Point (°C): -20 Freezing point (°C): 6 Specific Gravity (20/20): 0.78

Formula: C₆H₁₂
Boiling Pt: 81 °C
Melting Pt: 7 °C
Density: 0.78 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR