Eucalyptol ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane

C0542-25ML
TCC0542-25MLEA 180 USD
TCC0542-25ML
Eucalyptol ≥99.0% (by GC)
Eucalyptol

[for Determination of o-Cresol] CAS Number: 470-82-6 MDL Number: MFCD00167977 Molecular Formula: C10H18O Molecular Weight: 154.25 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 176 Melting point (°C): 2 Flash Point (°C): 48 Specific Gravity (20/20): 0.92


Product Categories: Bioscience, Bicyclic Monoterpenes Other Product Names / Synonyms: 1,8-Epoxy-p-menthane, Eucalyptol

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 176 °C
Melting Pt: 2 °C
Density: 0.92 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00167977
CAS Number: 470-82-6
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41149715b83e603fd6ea3a8ffc18a3c254c18939&vid=41149715b83e603fd6ea3a8ffc18a3c254c18939&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR