trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
C0475-500G
C0475-25G
C0475-100G
TCC0475-500GEA
408
USD
TCC0475-500G
TCC0475-025G
TCC0475-100G
trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)
trans-1,4-Cyclohexanedicarboxylic acid
CAS Number: 619-82-9
MDL Number: MFCD00066203
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method:
Formula:
C₈H₁₂O₄ |
MDL Number:
MFCD00066203 CAS Number: 619-82-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...