Cyanoguanidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0454-500G C0454-25G
TCC0454-500GEA 25 USD
TCC0454-500G TCC0454-025G
Cyanoguanidine ≥98.0% (by titrimetric analysis)
Cyanoguanidine

CAS Number: 461-58-5 MDL Number: MFCD00008066 Molecular Formula: C2H4N4 Molecular Weight: 84.08 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 211


Product Categories: Materials Chemistry, Polymerization Initiators Other Product Names / Synonyms: Cyanoguanidine

Formula: (H₂N)₂CNCN
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: 211 °C
Density: 1.404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS Number: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=261cad1de94683447fa014a27abc95d12ae00084&vid=261cad1de94683447fa014a27abc95d12ae00084&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR