3-Hydroxypropiononitrile ≥98.0%

Supplier: TCI America

C0450-500ML C0450-25ML C0450-100ML
TCC0450-500MLEA 483 USD
TCC0450-500ML TCC0450-025ML TCC0450-100ML
3-Hydroxypropiononitrile ≥98.0%
3-Hydroxypropiononitrile

CAS Number: 109-78-4 MDL Number: MFCD00002826 Molecular Formula: C3H5NO Molecular Weight: 71.08 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 221 Flash Point (°C): 137 Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Secondary Battery Research Reagents Other Product Names / Synonyms: 2-Cyanoethanol, Hydracrylonitrile, 3-Hydroxypropionitrile

Formula: C₃H₅NO
Boiling Pt: 221 °C
Melting Pt: –46 °C
Density: 1.05 (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38a2b321fec732a7ef21a5a66a116b4ebf4466c6&vid=38a2b321fec732a7ef21a5a66a116b4ebf4466c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR