Curcumin 0,1% in ethanol 95%

Supplier: TCI America

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

C0435-500ML
TCC0435-500MLEA 140 USD
TCC0435-500ML
Curcumin 0,1% in ethanol 95%
Curcumin

CAS Number: 458-37-7
MDL Number: MFCD00008365
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 13

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Flash Pt: 13 °C
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27d3c58ff42632d01947809785b9643f61982828&vid=6a371b832f14061bef5dabbb7b7b854ca966c49d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR