Crystal violet

Supplier: TCI America
Danger

C0428-500G C0428-25G C0428-100G
TCC0428-500GEA 229 USD
TCC0428-500G TCC0428-25G TCC0428-100G
Crystal violet
Crystal Violet

CAS Number: 548-62-9
MDL Number: MFCD00011750
Molecular Formula: C25H30ClN3
Molecular Weight: 407.99
Form: Crystal
Color: Yellow Green
Melting point (°C): 211


Other Product Names / Synonyms: Basic Violet 3, Hexamethylpararosaniline Chloride

Formula: C₂₅H₃₀ClN₃
Melting Pt: 211 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adea2ae2a9521d7ed63b9f59db4f8ec1c5ff8867&vid=b4b460d56ac2a684726430c5bd1368faf6fa697b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR