Chondroitin sulfate sodium salt

Supplier: TCI America

C0335-100G
TCC0335-100GEA 268 USD
TCC0335-100G
Chondroitin sulfate sodium salt
Chondroitin sulfate sodium salt
MDL Number: MFCD00130792
CAS Number: 9082-07-9

Order Now

Specification Test Results

Form Crystal
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR