Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

C0331-500G C0331-25G
TCC0331-500GEA 127.7 USD
TCC0331-500G TCC0331-025G
Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)
Choline dihydrogen citrate

CAS Number: 77-91-8 MDL Number: MFCD00038734 Molecular Formula: C11H21NO8 Molecular Weight: 295.29 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 107

Formula: C₁₁H₂₁NO₈
Melting Pt: 107 °C
MDL Number: MFCD00038734
CAS Number: 77-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89fd284a92cbbb10496de63fbbffd32257d37397&vid=89fd284a92cbbb10496de63fbbffd32257d37397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR