Choline chloride ≥98.0%
Supplier: TCI America
Synonyms:
2-hydroxy-N,N,N-trimethylethanaminium chloride
C0329-25G
C0329-500G
TCC0329-025GEA
23
USD
TCC0329-025G
TCC0329-500G
Choline chloride ≥98.0%
Choline chloride
CAS Number: 67-48-1
MDL Number: MFCD00011721
Molecular Formula: C5H14ClNO
Molecular Weight: 139.62
Purity/Analysis Method:
Product Categories: Bioscience, Other Plant Hormones, Vitamin Related Compounds
Other Product Names / Synonyms: (2-Hydroxyethyl)trimethylammonium Chloride
Formula:
HOH₂CCH₂N(CH₃)₃Cl MW: 139.62 g/mol Melting Pt: 302 °C Density: ∼1.205 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011721 CAS Number: 67-48-1 EINECS: 200-655-4 Merck Index: 13,02226 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...