1-Chloronaphthalene ≥85.0%

Supplier: TCI America
Warning

C0212-25G C0212-500G C0212-100G
TCC0212-025GEA 20.15 USD
TCC0212-025G TCC0212-500G TCC0212-100G
1-Chloronaphthalene ≥85.0%
1-Chloronaphthalene

CAS Number: 90-13-1 MDL Number: MFCD00003874 Molecular Formula: C10H7Cl Molecular Weight: 162.62 Purity/Analysis Method: 85.0% (GC) Form: Clear Liquid Boiling point (°C): 263 Melting point (°C): -20 Flash Point (°C): 121 Specific Gravity (20/20): 1.20

Formula: C₁₀H₇Cl
Boiling Pt: 263 °C
Melting Pt: –20 °C
Density: 1.20 (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 2810
ADR: 6.1,III
Merck Index: 13,02168
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da25412a8f7eb954e2111451072883555ce1689e&vid=da25412a8f7eb954e2111451072883555ce1689e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR